missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AP3D1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15961665
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15961665 100 μg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15961665 Supplier Invitrogen™ Supplier No. PA573013

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The AP3 amino acid sequence used as immunogen for this antibody is 100% homologous in human, chimpanzee, orangutan, new world monkey, rhesus monkey, and 63% homologous in panda, carolina anole, opossum, and 58% homologous in taipan, zebra finch, horse, dog, pig and 53% homologous in cow, rat, mouse.

AP3D1 is a subunit of the AP3 adaptor-like complex, which is not associated with clathrin. The AP3D1 subunit is implicated in intracellular biogenesis and trafficking of pigment granules and possibly platelet dense granules and neurotransmitter vesicles.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AP3D1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA and 0.05% sodium azide
Gene Ap3d1
Gene Accession No. O14617
Gene Alias AA407035; adapter-related protein complex 3 subunit delta-1; adaptin, delta; adaptor related protein complex 3 delta 1 subunit; adaptor related protein complex 3 subunit delta 1; adaptor related protein complex 3, delta 1 subunit; adaptor-related protein complex 3 subunit delta-1; adaptor-related protein complex 3, delta 1 subunit; adaptor-related protein complex 3, delta subunit; adaptor-related protein complex AP-3, delta subunit; ADTD; ap3; AP-3 complex delta subunit, partial CDS; AP-3 complex subunit delta; AP-3 complex subunit delta-1; Ap3d; Ap3d1; Bolvr; delta adaptin; delta-adaptin; delta-adaptin, partial CDS; hBLVR; mBLVR1; mh; mocha; PRO0039; subunit of putative vesicle coat adaptor complex AP-3
Gene Symbols Ap3d1
Host Species Rabbit
Immunogen A portion of amino acids 530-580 of human AP3 Delta.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8943
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.