missing translation for 'onlineSavingsMsg'
Learn More

Apigenin 7-Glucuronide, TRC

Product Code. 30011502
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011502 5 mg 5mg
30012060 1 mg 1mg
30006966 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011502 Supplier Toronto Research Chemicals Supplier No. TRCA7265055MG
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29741-09-1
Molecular Formula C21H18O11
Molecular Weight (g/mol) 446.36
Synonym β-D-5-Hydroxy-2-(p-hydroxyphenyl)-4-oxo-4H-1-benzopyran-7-yl-glucopyranosiduronic Acid, Apigenin 7-O-Glucuronide, Apigenin 7-O-β-D-Glucuronide, Apigenin 7-O-β-D-Glucuronopyranoside, Apigenin 7-O-β-Glucuronide, Apigenin 7-Glucuronide, Apigenin 7-β-Glucuronide, Apigenin-7-O-β-D Glucuronoside, Scutellarin A
IUPAC Name (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-[5-hydroxy-2-(4-hydroxyphenyl)-4-oxochromen-7-yl]oxyoxane-2-carboxylic acid
SMILES O[C@H]1[C@H](Oc2cc(O)c3C(=O)C=C(Oc3c2)c4ccc(O)cc4)O[C@@H]([C@@H](O)[C@@H]1O)C(=O)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C21H18O11/c22-9-3-1-8(2-4-9)13-7-12(24)15-11(23)5-10(6-14(15)31-13)30-21-18(27)16(25)17(26)19(32-21)20(28)29/h1-7,16-19,21-23,25-27H,(H,28,29)/t16-,17-,18+,19-,21+/m0/s1
Formula Weight 446.0849
Chemical Name or Material Apigenin 7-Glucuronide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.