missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Apolipoprotein L6 Polyclonal Antibody

Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.
This gene is a member of the apolipoprotein L gene family. The encoded protein is found in the cytoplasm, where it may affect the movement of lipids or allow the binding of lipids to organelles.
Specifications
Specifications
| Antigen | Apolipoprotein L6 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | APOL6 |
| Gene Accession No. | Q9BWW8 |
| Gene Alias | APOL6; apolipoprotein L, 6; Apolipoprotein L6; apolipoprotein L-VI; APOLVI; APOL-VI; DKFZp667M075; FLJ38562; FLJ90164; MGC57495; UNQ3095/PRO21341 |
| Gene Symbols | APOL6 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?