missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ ARHGAP9 Polyclonal Antibody
GREENER_CHOICE

Produktkode 17894730
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
100 μL
Förpackningsstorlek
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkode Quantity unitSize
17894730 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Denne vare kan ikke returneres. Se returpolitik
Produktkode 17894730 missing translation for 'mfr' Invitrogen™ missing translation for 'supplierNo' PA5140218

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolitik

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: NPGSMEGTQT LKRNNDVLQP QAKGFRSDTG TPEPLDPQGS LSLSQRTSQL DPPALQAPRP LPQLLDDPHE VEKSGLLNMT KIAQGGRKLR KNW Highest antigen sequence indentity to the following orthologs: Mouse - 59%, Rat - 43%.

This gene encodes a member of the Rho-GAP family of GTPase activating proteins. The protein has substantial GAP activity towards several Rho-family GTPases in vitro, converting them to an inactive GDP-bound state. It is implicated in regulating adhesion of hematopoietic cells to the extracellular matrix. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen ARHGAP9
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARHGAP9
Gene Accession No. Q9BRR9
Gene Alias 10C; ARHGAP9; RGL1; Rho GTPase activating protein 9; rho GTPase-activating protein 9; Rho-type GTPase-activating protein 9
Gene Symbols ARHGAP9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARHGAP9. Recombinant protein control fragment (Product #RP-100932).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64333
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.