missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arrestin 3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-69084
This item is not returnable.
View return policy
Description
Arrestin 3 Polyclonal specifically detects Arrestin 3 in Human samples. It is validated for Western Blot.
Specifications
| Arrestin 3 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| ARR3 | |
| Synthetic peptides corresponding to ARR3 (arrestin 3, retinal (X-arrestin)) The peptide sequence was selected from the C terminal of ARR3. Peptide sequence DVGVELPLVLIHPKPSHEAASSEDIVIEEFTRKGEEESQKAVEAEGDEGS. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| arrestin 3, retinal (X-arrestin), arrestin 4, arrestin-C, ARRXcArr, CAR, C-arrestin, Cone arrestin, Retinal cone arrestin-3, X-arrestin | |
| Rabbit | |
| 43 kDa | |
| 100 μL | |
| Cell Biology, Cellular Markers, GPCR, Neuroscience, Neurotransmission, Sensory Systems, Signal Transduction, Virology Bacteria and Parasites, Vision | |
| 407 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction