missing translation for 'onlineSavingsMsg'
Learn More

Arrestin 3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

483.00€

Specifications

Antigen Arrestin 3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18295001
View Documents
Novus Biologicals
NBP1-69084
100 μL
483.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Arrestin 3 Polyclonal specifically detects Arrestin 3 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Arrestin 3
Polyclonal
Rabbit
Cell Biology, Cellular Markers, GPCR, Neuroscience, Neurotransmission, Sensory Systems, Signal Transduction, Virology Bacteria and Parasites, Vision
arrestin 3, retinal (X-arrestin), arrestin 4, arrestin-C, ARRXcArr, CAR, C-arrestin, Cone arrestin, Retinal cone arrestin-3, X-arrestin
ARR3
IgG
43 kDa
Western Blot
Unconjugated
RUO
Human
407
Synthetic peptides corresponding to ARR3 (arrestin 3, retinal (X-arrestin)) The peptide sequence was selected from the C terminal of ARR3. Peptide sequence DVGVELPLVLIHPKPSHEAASSEDIVIEEFTRKGEEESQKAVEAEGDEGS.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.