Learn More
ATG7 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10088-2-AP-150UL
Description
The ATG7 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ATG7. This antibody recognizes human, mouse antigen. The ATG7 antibody has been validated for the following applications: IF, IHC, IP, WB, ELISA analysis.
Specifications
| ATG7 | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 10533 | |
| IgG |
| Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, ELISA | |
| 333 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse |