missing translation for 'onlineSavingsMsg'
Learn More

ATG7 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00€ - 410.00€

Specifications

Antigen ATG7
Concentration 333 μg/mL
Applications Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16856243
View Documents
Proteintech
10088-2-AP-20UL
20 μL
145.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16846243
View Documents
Proteintech
10088-2-AP-150UL
150 μL
410.00€
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The ATG7 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ATG7. This antibody recognizes human, mouse antigen. The ATG7 antibody has been validated for the following applications: IF, IHC, IP, WB, ELISA analysis.

Specifications

Specifications

ATG7
Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, ELISA
Unconjugated
Human, Mouse
IgG
333 μg/mL
Polyclonal
Rabbit
10533
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.