missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP Citrate Lyase Antibody (3A5), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26530-50ul
This item is not returnable.
View return policy
Description
ATP Citrate Lyase Monoclonal antibody specifically detects ATP Citrate Lyase in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry
Specifications
| ATP Citrate Lyase | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry | |
| 3A5 | |
| Western Blot 1:500-1:5000, Flow Cytometry 1:20-1:200, ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
| ACLEC 2.3.3.8, ATP citrate lyase, ATP citrate synthase, ATP-citrate (pro-S-)-lyase, ATP-citrate synthase, ATPCL, Citrate cleavage enzyme, CLATP | |
| A synthesized peptide derived from Human ATP Citrate Lyase [UniProt P53396] | |
| 50 μL | |
| Phospho Specific | |
| 47 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction