missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ATP6V0D2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16531442
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16531442 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16531442 Supplier Invitrogen™ Supplier No. PA598618

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ATP6V0D2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

ATP6V0D2 is the subunit of the integral membrane V0 complex of vacuolar ATPase. Vacuolar ATPase is responsible for acidifying a variety of intracellular compartments in eukaryotic cells, thus providing most of the energy required for transport processes in the vacuolar system. ATP6V0D2 may play a role in coupling of proton transport and ATP hydrolysis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP6V0D2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 4.045 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ATP6V0D2
Gene Accession No. Q8N8Y2
Gene Alias 1620401A02Rik; AI324824; ATP6D2; Atp6v0d2; ATPase H+ transporting V0 subunit d2; ATPase, H+ transporting, lysosomal 38kDa, V0 subunit d2; ATPase, H+ transporting, lysosomal V0 subunit D2; ATPase, H+ transporting, V0 subunit D, isoform 2; FLJ38708; osteoclast-specific vacuolar ATP synthase; similar to ATPase, H+ transporting, V0 subunit D isoform 2; vacuolar proton pump subunit d 2; vacuolar proton-translocating ATPase d subunit d2; V-ATPase; V-ATPase subunit d 2; VMA6; V-type proton ATPase subunit d 2
Gene Symbols ATP6V0D2
Host Species Rabbit
Immunogen Recombinant Human V-type proton ATPase subunit d 2 protein (50-295AA).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 245972
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.