missing translation for 'onlineSavingsMsg'
Learn More

B-Raf Antibody [Janelia Fluor™ 549], Novus Biologicals™

Product Code. 30627020 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30627020 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30627020 Supplier Novus Biologicals Supplier No. AF3424I

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

B-Raf Polyclonal antibody specifically detects B-Raf in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen B-Raf
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias 94 kDa B-raf protein, B-Raf proto-oncogene serine/threonine-protein kinase (p94), B-RAF1, BRAF1MGC126806, EC 2.7.11.1, MGC138284, murine sarcoma viral (v-raf) oncogene homolog B1, NS7, p94, Proto-oncogene B-Raf, RAFB1, serine/threonine-protein kinase B-raf, v-raf murine sarcoma viral oncogene homolog B1FLJ95109
Host Species Goat
Immunogen E. coli-derived recombinant human B-Raf, Met1-His766, Accession # P15056
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 673
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.