missing translation for 'onlineSavingsMsg'
Learn More

B7-2/CD86 Antibody (AP-MAB0803) - Azide and BSA Free, Novus Biologicals™

Product Code. 18074297 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
100 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18074297 0.025 mg 25µg
18023537 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18074297 Supplier Novus Biologicals Supplier No. NBP2121820.025MG
  • ONLINE EXCLUSIVE PRICE
    218.00€
    205.80€ / 25µg
    Save 12.20€
    5% Off
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 2 publications

B7-2/CD86 Monoclonal specifically detects B7-2/CD86 in Mouse samples. It is validated for CyTOF, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen B7-2/CD86
Applications CyTOF, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone AP-MAB0803
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:100-1000, Immunohistochemistry 1:-300-2000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:-300-2000, CyTOF-ready
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Alias Activation B7-2 antigen, B70, B7-2, B7-2 antigen, B-lymphocyte activation antigen B7-2, BU63, CD28 antigen ligand 2, CD28LG2B7-2 antigen), CD86 antigen, CD86 molecule, CTLA-4 counter-receptor B7.2, FUN-1, LAB72, MGC34413, T-lymphocyte activation antigen CD86
Gene Symbols CD86
Host Species Rat
Immunogen Mouse CD86
Purification Method Protein G purified
Quantity 0.025 mg
Research Discipline Adaptive Immunity, Cell Cycle and Replication, Dendritic Cell Markers, Diabetes Research, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 942
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.