missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B7-H4 Antibody (188), Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-43266-0.025MG
This item is not returnable.
View return policy
Description
B7-H4 Monoclonal specifically detects B7-H4 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| B7-H4 | |
| Monoclonal | |
| Unconjugated | |
| Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
| VTCN1 | |
| The immunogen for this antibody was Purified B7H4. | |
| 0.025 mg | |
| Primary | |
| Mouse | |
| IgG2b κ |
| Flow Cytometry | |
| 188 | |
| Flow Cytometry 1:10-1:1000 | |
| B7h.5, B7-H4, B7H4T-cell costimulatory molecule B7x, B7S1VCTN1, B7XPRO1291, FLJ22418, Immune costimulatory protein B7-H4, Protein B7S1, T cell costimulatory molecule B7x, V-set domain containing T cell activation inhibitor 1, V-set domain-containing T-cell activation inhibitor 1 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| 79679 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction