missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B7-H4 Antibody (188), Novus Biologicals™
Rat Monoclonal Antibody
433.00€ - 589.00€
Specifications
| Antigen | B7-H4 |
|---|---|
| Clone | 188 |
| Dilution | Flow Cytometry 1:10-1:1000 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18204572
|
Novus Biologicals
NBP1-43266-0.025MG |
0.025 mg |
433.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18222675
|
Novus Biologicals
NBP1-43266-0.1MG |
0.1 mg |
589.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
B7-H4 Monoclonal specifically detects B7-H4 in Mouse samples. It is validated for Flow Cytometry.Specifications
| B7-H4 | |
| Flow Cytometry 1:10-1:1000 | |
| Monoclonal | |
| Rat | |
| Mouse | |
| B7h.5, B7-H4, B7H4T-cell costimulatory molecule B7x, B7S1VCTN1, B7XPRO1291, FLJ22418, Immune costimulatory protein B7-H4, Protein B7S1, T cell costimulatory molecule B7x, V-set domain containing T cell activation inhibitor 1, V-set domain-containing T-cell activation inhibitor 1 | |
| VTCN1 | |
| IgG2b κ | |
| Affinity Purified |
| 188 | |
| Flow Cytometry | |
| Unconjugated | |
| RUO | |
| Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
| 79679 | |
| The immunogen for this antibody was Purified B7H4. | |
| Primary | |
| Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title