missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAFFR/TNFRSF13C Antibody (BAFFR/1558), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08868AF700
This item is not returnable.
View return policy
Description
BAFFR/TNFRSF13C Monoclonal antibody specifically detects BAFFR/TNFRSF13C in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin)
Specifications
| BAFFR/TNFRSF13C | |
| Monoclonal | |
| Alexa Fluor 700 | |
| BAFFR;BAFF-R;BROMIX;CD268;CVID4;prolixin;Tumor necrosis factor receptor superfamily member 13C | |
| Recombinant full-length human BAFFR/TNFRSF13C protein (Uniprot: Q96RJ3) | |
| 100 μg | |
| Biologically Active Proteins, Cytokine Research, Immunology, Lipid and Metabolism | |
| 115650 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry (Paraffin) | |
| BAFFR/1558 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?