missing translation for 'onlineSavingsMsg'
Learn More

Bayogenin, TRC

Product Code. 30003893
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
5 mg
Unit Size:
0.5mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003893 1 mg 1mg
30010259 5 mg 5mg
30010419 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003893 Supplier Toronto Research Chemicals Supplier No. TRCB1190181MG
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6989-24-8
Molecular Formula C30H48O5
Molecular Weight (g/mol) 488.7
Synonym Olean-12-en-28-oic acid, 2,3,23-trihydroxy-, (2β,3β,4α)-, Olean-12-en-28-oic acid, 2β,3β,23-trihydroxy- (7CI,8CI), 2β,3β,23α-Trihydroxy-12-oleanen-28-oic acid, Bayogenin, Bayogenine
IUPAC Name (4aS,6aR,6aS,6bR,8aR,9R,10R,11S,12aR,14bS)-10,11-dihydroxy-9-(hydroxymethyl)-2,2,6a,6b,9,12a-hexamethyl-1,3,4,5,6,6a,7,8,8a,10,11,12,13,14b-tetradecahydropicene-4a-carboxylic acid
SMILES CC1(C)CC[C@@]2(CC[C@]3(C)C(=CC[C@@H]4[C@@]5(C)C[C@H](O)[C@H](O)[C@@](C)(CO)[C@@H]5CC[C@@]34C)[C@@H]2C1)C(=O)O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C30H48O5/c1-25(2)11-13-30(24(34)35)14-12-28(5)18(19(30)15-25)7-8-22-26(3)16-20(32)23(33)27(4,17-31)21(26)9-10-29(22,28)6/h7,19-23,31-33H,8-17H2,1-6H3,(H,34,35)/t19-,20-,21+,22+,23-,26-,27-,28+,29+,30-/m0/s1
Formula Weight 488.3502
Chemical Name or Material Bayogenin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.