missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCL2L12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00313-50ul
This item is not returnable.
View return policy
Description
BCL2L12 Polyclonal antibody specifically detects BCL2L12 in Human samples. It is validated for ELISA
Specifications
| BCL2L12 | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| Bcl-2 related proline-rich protein, bcl2-L-12, BCL2-like 12 (proline rich), bcl-2-like protein 12, Bcl-2-related proline-rich protein, BPR, MGC120313, MGC120314, MGC120315 | |
| Produced in rabbits immunized with purified, recombinant Human BCL2L12 (Uniprot#: Q9HB09-1; Pro147-Lys227). | |
| 50 μg | |
| Apoptosis | |
| 83596 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction