missing translation for 'onlineSavingsMsg'
Learn More

Benzyl-2,3,4,5,6-d5-dimethyl-n-dodecylammonium Chloride, TRC

Product Code. 30454021
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30454021 100 mg 100mg
30449560 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30454021 Supplier Toronto Research Chemicals Supplier No. TRCZ343001100MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzyl-2,3,4,5,6-d5-dimethyl-n-dodecylammonium Chloride
Molecular Formula C21H33D5ClN
Quantity 100 mg
InChI Formula InChI=1S/C21H38N.ClH/c1-4-5-6-7-8-9-10-11-12-16-19-22(2,3)20-21-17-14-13-15-18-21;/h13-15,17-18H,4-12,16,19-20H2,1-3H3;1H/q+1;/p-1/i13D,14D,15D,17D,18D;
SMILES [2H]C1=C([2H])C([2H])=C([2H])C([2H])=C1C[N+](C)(C)CCCCCCCCCCCC.[Cl-]
IUPAC Name N,N-dimethyl-N-((phenyl-d5)methyl)dodecan-1-aminium chloride
Molecular Weight (g/mol) 345.02
Formula Weight 344.3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.