missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Alcohol-OD, CDN

Product Code. 30314741
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30314741 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30314741 Supplier CDN Supplier No. CDND72405G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzyl Alcohol-O-D
CAS 14915-25-4
Molecular Formula C7 D H7 O
Quantity 5 g
Synonym Benzenemethanol-d (9 CI, ACI), Benzyl alcohol-d (7 CI, 8 CI), Benzyl alcohol-d1, Benzyl Alcohol-O-D, Benzyl Alcohol-OD
InChI Formula InChI=1 S/C7H8O/c8-6-7-4-2-1-3-5-7/h1-5,8 H,6H2/i8D
SMILES [2 H]OCc1ccccc1
IUPAC Name deuteriooxymethylbenzene
Molecular Weight (g/mol) 109.144
Formula Weight 109.0638 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.