missing translation for 'onlineSavingsMsg'
Learn More

Benzyl methacrylate, 98%, stab. with ca 50-100ppm 4-methoxyphenol

Product Code. 11313026
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11313026 50 g 50g
11323026 250 g 250g
11333026 1000 g 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11313026 Supplier Thermo Scientific Alfa Aesar Supplier No. L10840.18
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2495-37-6 | C11H12O2 | 176.215 g/mol

Benzyl methacrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl methacrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to light and heat. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2495-37-6
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.215
MDL Number MFCD00053684
InChI Key AOJOEFVRHOZDFN-UHFFFAOYSA-N
Synonym benzyl methacrylate, 2-propenoic acid, 2-methyl-, phenylmethyl ester, benzylmethacrylate, methacrylic acid benzyl ester, methacrylic acid, benzyl ester, unii-z3248k2ssm, phenylmethyl 2-methylprop-2-enoate, poly benzylmethacrylate, bzma, methacrylic acid benzyl
PubChem CID 17236
IUPAC Name benzyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCC1=CC=CC=C1

Specifications

Density 1.04
Boiling Point 231°C to 233°C
Flash Point 107°C (225°F)
Refractive Index 1.514
Quantity 50 g
Beilstein 2045924
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 176.22
Percent Purity 98%
Chemical Name or Material Benzyl methacrylate, stabilized with ≈50 to 100ppm 4-methoxyphenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.