missing translation for 'onlineSavingsMsg'
Learn More

Benzyl methyl malonate, 95%

Product Code. 15475358
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15475358 5 g 5g
15485358 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15475358 Supplier Thermo Scientific Alfa Aesar Supplier No. H59140.06
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 52267-39-7 | C11H12O4 | 208.21 g/mol

It is employed as a intermediate for pharmaceutical and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceutical and organic synthesis.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52267-39-7
Molecular Formula C11H12O4
Molecular Weight (g/mol) 208.21
MDL Number MFCD00008461
InChI Key IAUZDBFOEWAQFE-UHFFFAOYSA-N
Synonym benzyl methyl malonate, 1-benzyl 3-methyl propanedioate, malonic acid 1-benzyl 3-methyl ester, propanedioic acid, methyl phenylmethyl ester, benzylmethyl malonate, methyl benzyl malonate, acmc-1b0zd, benzyl methyl propanedioate, ksc493e1f
PubChem CID 572135
IUPAC Name 3-O-benzyl 1-O-methyl propanedioate
SMILES COC(=O)CC(=O)OCC1=CC=CC=C1

Specifications

Density 1.15
Boiling Point 125°C (0.5 mmHg)
Flash Point >113°C (235°F)
Linear Formula C6H5CH2OCOCH2COOCH3
Refractive Index 1.502
Quantity 5 g
Beilstein 2529986
Solubility Information Slightly soluble in water.
Formula Weight 208.21
Percent Purity 95%
Chemical Name or Material Benzyl methyl malonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.