missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Phenylephrone Hydrochloride, TRC

Product Code. 17960605
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17960605 50 mg 50mg
17960365 10 mg 10mg
17904955 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17960605 Supplier Toronto Research Chemicals Supplier No. TRCB28863550MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71786-67-9
Molecular Formula C16 H17 N O2 . Cl H
Molecular Weight (g/mol) 291.77
Synonym 2-(Benzyl-methylamino)-1-(3-hydroxyphenyl)ethanone Hydrochloride, Phenylephrine Hydrochloride Imp. E (EP) as Hydrochloride, Benzylphenylephrone Hydrochloride, 2-(N-Benzyl-N-methylamino)-1-(3-hydroxyphenyl)ethanone hydrochloride
IUPAC Name 2-[benzyl(methyl)amino]-1-(3-hydroxyphenyl)ethanone;hydrochloride
SMILES Cl.CN(CC(=O)c1cccc(O)c1)Cc2ccccc2

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C16H17NO2.ClH/c1-17(11-13-6-3-2-4-7-13)12-16(19)14-8-5-9-15(18)10-14;/h2-10,18H,11-12H2,1H3;1H
Formula Weight 291.1026
Percent Purity >95%
Chemical Name or Material 2-(Benzyl-methylamino)-1-(3-hydroxyphenyl)ethanone Hydrochloride (Benzylphenylephrone Hydrochloride)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.