Learn More
Bestrophin 1 Antibody (E6-6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 42 publications
Brand:  Novus Biologicals NB300-164
 View more versions of this product
Product Code. 18740014
  • 590.00€ / 0.10mL
Description
Bestrophin 1 Monoclonal specifically detects Bestrophin 1 in Human, Mouse, Porcine, Canine, Primate, Rat (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Proximity Ligation Assay, Dual RNAscope ISH-IHC.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), In situ PLA
Pig, Primate, Rat (Negative)
Ascites
Monoclonal
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.