missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bestrophin 1 Antibody (E6-6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 42 publications
308.00€ - 590.00€
Specifications
| Applications | Western Blot, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
|---|---|
| Classification | Monoclonal |
| Form | Ascites |
| Target Species | Mouse, Pig |
| Isotype | IgG1 κ |
| Product Code | Brand | dimQuantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | dimQuantity | Price | Quantity & Availability | |||||
|
18731384
|
Novus Biologicals
NB300-164SS |
308.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
|
18740014
|
Novus Biologicals
NB300-164 |
590.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Bestrophin 1 Monoclonal specifically detects Bestrophin 1 in Human, Mouse, Porcine, Canine, Primate, Rat (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Proximity Ligation Assay, Dual RNAscope ISH-IHC.Specifications
| Western Blot, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Ascites | |
| IgG1 κ |
| Monoclonal | |
| Mouse, Pig |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title