missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beta Lactoglobulin B Antibody - BSA Free, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals NB100-62383-0.1ML
This item is not returnable.
View return policy
Description
Beta Lactoglobulin B Polyclonal specifically detects Beta Lactoglobulin B in Bovine samples. It is validated for ELISA.
Specifications
| Beta Lactoglobulin B | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.09% Sodium Azide | |
| beta-lactoglobulin pseudogene | |
| Sheep | |
| IgG purified | |
| RUO | |
| 138159 | |
| Bovine | |
| Purified |
| ELISA | |
| 5 mg/mL | |
| ELISA 1:100-1:2000 | |
| P02754 | |
| LOC138159 | |
| Beta-lactoglobulin B from milk | |
| 0.1 mL | |
| Primary | |
| This product recognizes bovine beta-lactoglobulin B, a component of whey thought to be involved in retinol transport. This antibody also demonstrates some cross reactivity with beta-lactoglobulin A. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction