missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beta Lactoglobulin B Antibody - BSA Free, Novus Biologicals™
Sheep Polyclonal Antibody
204.00€ - 467.00€
Specifications
| Antigen | Beta Lactoglobulin B |
|---|---|
| Concentration | 5 mg/mL |
| Dilution | ELISA 1:100-1:2000 |
| Applications | ELISA |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18097863
|
Novus Biologicals
NB100-62383-0.1ML |
0.1 mL |
204.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18063104
|
Novus Biologicals
NB100-62383 |
0.5 mL |
467.00€
0.50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Beta Lactoglobulin B Polyclonal specifically detects Beta Lactoglobulin B in Bovine samples. It is validated for ELISA.Specifications
| Beta Lactoglobulin B | |
| ELISA 1:100-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 0.09% Sodium Azide | |
| beta-lactoglobulin pseudogene | |
| LOC138159 | |
| IgG | |
| IgG purified | |
| This product recognizes bovine beta-lactoglobulin B, a component of whey thought to be involved in retinol transport. This antibody also demonstrates some cross reactivity with beta-lactoglobulin A. |
| 5 mg/mL | |
| ELISA | |
| Unconjugated | |
| Sheep | |
| Bovine | |
| P02754 | |
| 138159 | |
| Beta-lactoglobulin B from milk | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title