Learn More
Beta-Zearalenol-d7, TRC
Specifications
Specifications
| Chemical Name or Material | beta-Zearalenol-d7 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C18H17D7O5 |
| Quantity | 1 mg |
| Synonym | (3S,7S,11E)-3,4,5,6,7,8,9,10-Octahydro-7,14,16-trihydroxy-3-methyl-1H-2-benzoxacyclotetradecin-1-one-d7, (-)-β-Zearalenol-d7, β-trans-Zearalenol-d7 |
| InChI Formula | InChI=1S/C18H24O5/c1-12-6-5-9-14(19)8-4-2-3-7-13-10-15(20)11-16(21)17(13)18(22)23-12/h3,7,10-12,14,19-21H,2,4-6,8-9H2,1H3/b7-3+/t12-,14-/m0/s1/i8D2,9D2,10D,11D,14D |
| SMILES | O=C1O[C@@H](C)CCC([2H])([2H])[C@@]([2H])(O)C([2H])([2H])CC/C=C/C2=C([2H])C(O)=C([2H])C(O)=C21 |
| Molecular Weight (g/mol) | 327.42 |
| Percent Purity | >95 |