missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Betula pendula BETVIA Chimeric Recombinant Rabbit Monoclonal Antibody (BV16)

Description
Specificity: This antibody binds to Bet v 1.
May be a general steroid carrier protein.
Specifications
Specifications
| Antigen | Betula pendula BETVIA Chimeric |
| Applications | ELISA, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | BV16 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% ProClin 300 |
| Gene | ITGA4 |
| Gene Accession No. | P13612 |
| Gene Alias | allergen Bet v 1-A; Allergen Bet v I-A; BETVI; Major pollen allergen Bet v 1-A |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?