missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biotin (Vitamin B7 or Vitamin H) Antibody (Hyb-8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-32811-0.1mg
This item is not returnable.
View return policy
Description
Biotin (Vitamin B7 or Vitamin H) Monoclonal antibody specifically detects Biotin (Vitamin B7 or Vitamin H) in Nonspecies specific samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), In-situ Hybridization
Specifications
| Biotin (Vitamin B7 or Vitamin H) | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 0.5-2 ug/mL, Flow Cytometry 0.5-1 ug/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 0.5-1 ug/mL, Immunohistochemistry-Paraffin 0.5-1 ug/mL, In-situ Hybridization | |
| 58-85-5, coenzyme R, d-biotin, Vitamin B7, vitamin H | |
| Biotinylated sheep immunoglobulin | |
| 0.1 mg | |
| Primary | |
| Store at 4C. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), In Situ Hybridization (ISH) | |
| Hyb-8 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction