missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biotin (Vitamin B7 or Vitamin H) Antibody (Hyb-8), Novus Biologicals™
Mouse Monoclonal Antibody
290.00€ - 504.00€
Specifications
| Antigen | Biotin (Vitamin B7 or Vitamin H) |
|---|---|
| Clone | Hyb-8 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 0.5-2 ug/mL, Flow Cytometry 0.5-1 ug/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 0.5-1 ug/mL, Immunohistochemistry-Paraffin 0.5-1 ug/mL, In-situ Hybridization |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), In Situ Hybridization (ISH) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18440232
|
Novus Biologicals
NBP2-32811-0.1mg |
0.1 mg |
504.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18463452
|
Novus Biologicals
NBP2-32811-0.02mg |
0.02 mg |
290.00€
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Biotin (Vitamin B7 or Vitamin H) Monoclonal antibody specifically detects Biotin (Vitamin B7 or Vitamin H) in Nonspecies specific samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), In-situ HybridizationSpecifications
| Biotin (Vitamin B7 or Vitamin H) | |
| 0.2 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), In Situ Hybridization (ISH) | |
| Unconjugated | |
| Mouse | |
| Non-species specific | |
| 58-85-5, coenzyme R, d-biotin, Vitamin B7, vitamin H | |
| IgG1 κ | |
| Protein A or G purified |
| Hyb-8 | |
| Western Blot 0.5-2 ug/mL, Flow Cytometry 0.5-1 ug/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 0.5-1 ug/mL, Immunohistochemistry-Paraffin 0.5-1 ug/mL, In-situ Hybridization | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| Biotinylated sheep immunoglobulin | |
| Primary | |
| Store at 4C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title