missing translation for 'onlineSavingsMsg'
Learn More

Bis(pentamethylcyclopentadienyl)hafnium(IV) dichloride, 98+%

Product Code. 15417635
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15417635 1 g 1g
15427635 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15417635 Supplier Thermo Scientific Alfa Aesar Supplier No. H26912.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 85959-83-7 | C20H30Cl2Hf | 519.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85959-83-7
Molecular Formula C20H30Cl2Hf
Molecular Weight (g/mol) 519.85
MDL Number MFCD00058887
InChI Key YPRLSLCBPOTFGE-UHFFFAOYSA-L
Synonym 1,2,3,5,5-pentamethylcyclopenta-1,3-diene;dichloride, hafnium,dichlorobis 1,2,3,4,5-h-1,2,3,4,5-pentamethyl-2,4-cyclopentadien-1-yl
PubChem CID 21908586
IUPAC Name hafnium(4+);1,2,3,5,5-pentamethylcyclopenta-1,3-diene;dichloride
SMILES CC1=[C-]C(C(=C1C)C)(C)C.CC1=[C-]C(C(=C1C)C)(C)C.[Cl-].[Cl-].[Hf+4]

Specifications

Melting Point 300°C
Quantity 1 g
Formula Weight 519.86
Percent Purity ≥98%
Chemical Name or Material Bis(pentamethylcyclopentadienyl)hafnium(IV) dichloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.