missing translation for 'onlineSavingsMsg'
Learn More

Bismuth rod, Puratronic™, 99.9999% (metals basis), Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar 047229.KF

Quantity 1 pc.

 View more versions of this product

Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 15474125

  • 691.00€ / Pack of 1
 Restricted Item
In Stock
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

7440-69-9
208.98
JCXGWMGPZLAOME-UHFFFAOYSA-N
5359367
bismuth
Bi
MFCD00134033
bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod
CHEBI:33301
[Bi]

Specifications

7440-69-9
Odorless
Bi
bismuth, elemental, unii-u015tt5i8h, powder, iii ion, shot, elongated, rod, 12.7mm 0.5in dia, atom, needles, pieces, rod
[Bi]
208.98
CHEBI:33301
Puratronic™
Bismuth rod, 10mm (0.39 in.) dia. x 145mm (5.71 in.) long
1 pc.
≈7.5g/cm
MFCD00134033
JCXGWMGPZLAOME-UHFFFAOYSA-N
bismuth
5359367
99.9999%
(metals basis)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.