Learn More
Blooms Syndrome Protein Blm Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand:  Novus Biologicals NBP1-89929
 View more versions of this product
Product Code. 18777483
  • 554.00€ / 0.10mL
Description
Blooms Syndrome Protein Blm Polyclonal specifically detects Blooms Syndrome Protein Blm in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Immunocytochemistry, Immunofluorescence
Rabbit
RUO
Polyclonal
0.1 mL
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.