missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Blooms Syndrome Protein Blm Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
369.00€ - 554.00€
Specifications
| Antigen | Blooms Syndrome Protein Blm |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18422462
|
Novus Biologicals
NBP1-89929-25ul |
25 μL |
369.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18777483
|
Novus Biologicals
NBP1-89929 |
0.1 mL |
554.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Blooms Syndrome Protein Blm Polyclonal specifically detects Blooms Syndrome Protein Blm in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Blooms Syndrome Protein Blm | |
| Polyclonal | |
| Rabbit | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Genes Sensitive to DNA Damaging Agents, Stem Cell Markers, Tumor Suppressors | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 641 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:STLKDLDTSDRKEDVLSTSKDLLSKPEKMSMQELNPETSTDCDARQISLQQQLIHVMEHICKLIDTIPDDKLKLLDCGNELLQQRNIRRKLLTEVDFNKSDASLLGSLWRYRPDSLDGPMEGDSCPTGNSMKELNFSHLPSNSV | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| Bloom syndrome, Bloom syndrome, RecQ helicase-like, BSBloom syndrome protein, DNA helicase, RecQ-like type 2, EC 3.6.1, EC 3.6.4.12, MGC126616, MGC131618, RecQ protein-like 3, RecQ2, RECQ2MGC131620, RECQL3RECQL2 | |
| BLM | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title