missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BMP-8A Polyclonal Antibody

Description
This antibody detects endogenous protein at a molecular weight of 45 kDa. It may cross-react with BMP-8B protein. Purity is >95% by SDS-PAGE.
Bone morphogenic proteins (BMPs) are members of the TGF beta superfamily. BMPs are involved in the induction of cartilage and bone formation.BMP-8 is thought to be involved in early development, as detectable expression has not been found in adult organs. Two BMP-8 proteins exist, namely BMP-8A and BMP-8B (also designated OP-2), and are encoded by two distinct genes.
Specifications
Specifications
| Antigen | BMP-8A |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | BMP8A |
| Gene Accession No. | P34821, Q7Z5Y6 |
| Gene Alias | BMP; Bmp7r1; Bmp8; Bmp-8; BMP8A; BMP-8A; BMP8B; BMP-8B; Bone morphogenetic protein; Bone morphogenetic protein 8A; OP2; OP-2; Osteogenic protein 2 |
| Gene Symbols | BMP8A |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?