missing translation for 'onlineSavingsMsg'
Learn More

BPIFB1 Antibody (1023938) [mFluor Violet 450 SE], Novus Biologicals™

Product Code. 30614201 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30614201 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30614201 Supplier Novus Biologicals Supplier No. FAB10195MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

BPIFB1 Monoclonal antibody specifically detects BPIFB1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen BPIFB1
Applications Western Blot
Classification Monoclonal
Clone 1023938
Conjugate mFluor Violet 450 SE
Formulation 50mM Sodium Borate
Gene Alias bA49G10.6, chromosome 20 open reading frame 114, dJ1187J4.1, long palate, lung and nasal epithelium carcinoma associated 1, long palate, lung and nasal epithelium carcinoma-associated protein 1, LPLUNC1, MGC14597, VEMSGP, Von Ebner minor salivary gland protein
Host Species Mouse
Immunogen Human embryonic kidney cell, HEK293-derived human BPIFB1, Thr22-Gln484, Accession # Q8TDL5
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92747
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.