Learn More
BRD4 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28486-1-AP-20UL
Description
The BRD4 antibody from Proteintech is a rabbit polyclonal antibody to a peptide of human BRD4. This antibody recognizes human antigen. The BRD4 antibody has been validated for the following applications: IP, WB, ELISA analysis.
Specifications
| BRD4 | |
| Polyclonal | |
| Rabbit | |
| Primary | |
| Human |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| 20 μL | |
| 23476 | |
| IgG |