missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRD4 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | BRD4 |
|---|---|
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17581366
|
Proteintech
28486-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17571366
|
Proteintech
28486-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The BRD4 antibody from Proteintech is a rabbit polyclonal antibody to a peptide of human BRD4. This antibody recognizes human antigen. The BRD4 antibody has been validated for the following applications: IP, WB, ELISA analysis.
Specifications
| BRD4 | |
| Polyclonal | |
| Rabbit | |
| 23476 | |
| Primary |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Human | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title