missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1qR1/CD93 Antibody (R139) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-43378-0.1MG
This item is not returnable.
View return policy
Description
C1qR1/CD93 Monoclonal specifically detects C1qR1/CD93 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunoprecipitation.
Specifications
| C1q R1/CD93 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunoprecipitation 1:10-1:500 | |
| C1q receptor 1, C1q/MBL/SPA receptor, C1qR(P), C1qRp, CD93 antigenC1qR, CD93 molecule, CDw93C1QR1, Complement component 1 q subcomponent receptor 1, complement component 1, q subcomponent, receptor 1, complement component C1q receptor, dJ737E23.1, ECSM3, Matrix-remodeling-associated protein 4, matrix-remodelling associated 4, MXRA4 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 22918 | |
| Store at 4C. Do not freeze. |
| Western Blot, Flow Cytometry, ELISA, Immunoprecipitation | |
| R139 | |
| Unconjugated | |
| Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
| CD93 | |
| The immunogen for this antibody was CD93. | |
| 0.1 mg | |
| Primary | |
| Human | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction