missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1qR1/CD93 Antibody (R139) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
197.00€ - 517.00€
Specifications
| Antigen | C1q R1/CD93 |
|---|---|
| Clone | R139 |
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunoprecipitation 1:10-1:500 |
| Applications | Flow Cytometry, Western Blot, ELISA, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18283511
|
Novus Biologicals
NBP1-43378-25UG |
25 μg |
197.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18247371
|
Novus Biologicals
NBP1-43378-0.1MG |
0.1 mg |
517.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
C1qR1/CD93 Monoclonal specifically detects C1qR1/CD93 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunoprecipitation.Specifications
| C1q R1/CD93 | |
| 0.5 mg/mL | |
| Flow Cytometry, Western Blot, ELISA, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
| 22918 | |
| The immunogen for this antibody was CD93. | |
| Primary | |
| Store at 4C. Do not freeze. |
| R139 | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunoprecipitation 1:10-1:500 | |
| Monoclonal | |
| Mouse | |
| Human | |
| C1q receptor 1, C1q/MBL/SPA receptor, C1qR(P), C1qRp, CD93 antigenC1qR, CD93 molecule, CDw93C1QR1, Complement component 1 q subcomponent receptor 1, complement component 1, q subcomponent, receptor 1, complement component C1q receptor, dJ737E23.1, ECSM3, Matrix-remodeling-associated protein 4, matrix-remodelling associated 4, MXRA4 | |
| CD93 | |
| IgG2b | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title