missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CABP1 Monoclonal antibody specifically detects CABP1 in Human, Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | CABP1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | SR1754 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000 |
| Formulation | PBS, pH 7.4, 150mM NaCl, 50% glycerol. |
| Gene Alias | CaBP1, Calbrain, calcium binding protein 1, calcium binding protein 5, calcium-binding protein 1, caldendrin, HCALB_BR |
| Host Species | Rabbit |
| Immunogen | A synthesized peptide derived from human CABP1 (Uniprot #: Q9NZU7) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?