missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CACNB3 Polyclonal antibody specifically detects CACNB3 in Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | CACNB3 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | CAB3, CACNLB3, Calcium channel voltage-dependent subunit beta 3, calcium channel, voltage-dependent, beta 3 subunit, voltage-dependent L-type calcium channel subunit beta-3 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 355-484 of human CACNB3 (NP_000716.2).,, Sequence:, VYWRATHHPAPGPGLLGPPSAIPGLQNQQLLGERGEEHSPLERDSLMPSDEASESSRQAWTGSSQRSSRHLEEDYADAYQDLYQPHRQHTSGLPSANGHDPQDRLLAQDSEHNHSDRNWQRNRPWPKDSY |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?