missing translation for 'onlineSavingsMsg'
Learn More

Calretinin Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2D7A9, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66496-1-IG-150UL

 View more versions of this product

Product Code. 16858425

  • 410.00€ / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Calretinin is encoded by the CALB2 gene located on chromosome 16. It is a troponin C superfamily protein having six EF-hand structural motifs. The EF motif allows the protein to bind calcium, and function in calcium signaling, calcium buffering, and modulating neuronal excitability. Calretinin is expressed in the mesothelium, mast cells, some neural cells, hair follicles, and fat cells.
Specifications

Specifications

Calretinin
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P22676, P47728, Q08331
CALB2
Calretinin Fusion Protein Ag2924
150 μL
Primary
Human, Mouse, Pig, Rat
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
2D7A9
Unconjugated
CALB2
29 kDa calbindin, CAB29, CAL2, CALB2, calbindin 2, Calretinin, CR
Mouse
Protein G
RUO
100127479, 117059, 12308, 794
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.