missing translation for 'onlineSavingsMsg'
Learn More

Calretinin Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2D7A9, Proteintech

Mouse Monoclonal Antibody

145.00€ - 410.00€

Specifications

Antigen Calretinin
Clone 2D7A9
Concentration 1 mg/mL
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16868425
View Documents
Proteintech
66496-1-IG-20UL
20 μL
145.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16858425
View Documents
Proteintech
66496-1-IG-150UL
150 μL
410.00€
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Calretinin is encoded by the CALB2 gene located on chromosome 16. It is a troponin C superfamily protein having six EF-hand structural motifs. The EF motif allows the protein to bind calcium, and function in calcium signaling, calcium buffering, and modulating neuronal excitability. Calretinin is expressed in the mesothelium, mast cells, some neural cells, hair follicles, and fat cells.
Specifications

Specifications

Calretinin
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
29 kDa calbindin, CAB29, CAL2, CALB2, calbindin 2, Calretinin, CR
CALB2
IgG1
Protein G
Antibody
2D7A9
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Pig, Human, Mouse, Rat
P22676, P47728, Q08331
100127479, 117059, 12308, 794
Calretinin Fusion Protein Ag2924
Primary
-20°C
CALB2
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.