missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calretinin Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2D7A9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66496-1-IG-20UL
This item is not returnable.
View return policy
Description
Calretinin is encoded by the CALB2 gene located on chromosome 16. It is a troponin C superfamily protein having six EF-hand structural motifs. The EF motif allows the protein to bind calcium, and function in calcium signaling, calcium buffering, and modulating neuronal excitability. Calretinin is expressed in the mesothelium, mast cells, some neural cells, hair follicles, and fat cells.Specifications
| Calretinin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P22676, P47728, Q08331 | |
| CALB2 | |
| Calretinin Fusion Protein Ag2924 | |
| 20 μL | |
| Primary | |
| Pig, Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 2D7A9 | |
| Unconjugated | |
| CALB2 | |
| 29 kDa calbindin, CAB29, CAL2, CALB2, calbindin 2, Calretinin, CR | |
| Mouse | |
| Protein G | |
| RUO | |
| 100127479, 117059, 12308, 794 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction