missing translation for 'onlineSavingsMsg'
Learn More

Carbetocin, TRC

Product Code. 30006653
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006653 100 mg 100mg
30008153 10 mg 10mg
30011977 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006653 Supplier Toronto Research Chemicals Supplier No. TRCC175925100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37025-55-1
Synonym Glycinamide, N-(4-mercapto-1-oxobutyl)-O-methyl-L-tyrosyl-L-isoleucyl-L-glutaminyl-L-asparaginyl-L-cysteinyl-L-prolyl-L-leucyl-, cyclic (1→5)-thioether∣1-Carbaoxytocin, 1-butanoic acid-2-(O-methyl-L-tyrosine)- (9CI)∣1-Thia-4,7,10,13,16-pentaazacycloeicosane, cyclic peptide deriv.∣Carbetocin∣Deamino-2-O-methyltyrosine-1-carbaoxytocin∣Depotocin∣Duratocin∣Pabal∣[2-O-Methyltyrosine]-deamino-1-carba-oxytocin∣[Desamino-monocarba1,Tyr(methyl)2]-oxytocin

Specifications

Format Neat
Molecular Formula C45H69N11O12S
MDL Number MFCD01076600
Molecular Weight (g/mol) 988.16, 988.17
InChI Key SMMBGKXBHJTULT-CPWZVOFUSA-N
SMILES CC[C@H](C)[C@@H]1NC(=O)[C@H](CC2=CC=C(O)C=C2)N(C)C(=O)CCCSC[C@H](NC(=O)[C@H](CC(N)=O)NC(=O)[C@H](CCC(N)=O)NC1=O)C(=O)N1CCC[C@H]1C(=O)N[C@@H](CC(C)C)C(=O)NCC(N)=O
IUPAC Name (2S)-2-{[(2S)-1-[(3R,6S,9S,12S,15S)-12-[(2S)-butan-2-yl]-9-(2-carbamoylethyl)-6-(carbamoylmethyl)-15-[(4-hydroxyphenyl)methyl]-16-methyl-5,8,11,14,17-pentaoxo-1-thia-4,7,10,13,16-pentaazacycloicosane-3-carbonyl]pyrrolidin-2-yl]formamido}-N-(carbamoylmethyl)-4-methylpentanamide
Product Type Metabolite∣API
Quantity 100 mg
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.