missing translation for 'onlineSavingsMsg'
Learn More

Carbofuran D3 (N-methyl D3) 100 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30315985
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30315985 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30315985 Supplier Dr. Ehrenstorfer Supplier No. DREA11010100ME100
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Carbofuran-d3
CAS 1007459-98-4
Solvent or Matrix Methanol
Molecular Formula C12 2H3 H12 N O3
Quantity 1 mL
InChI Formula InChI=1S/C12H15NO3/c1-12(2)7-8-5-4-6-9(10(8)16-12)15-11(14)13-3/h4-6H,7H2,1-3H3,(H,13,14)/i3D3
SMILES [2H]C([2H])([2H])NC(=O)Oc1cccc2CC(C)(C)Oc12
IUPAC Name (2,2-dimethyl-3H-1-benzofuran-7-yl) N-(trideuteriomethyl)carbamate
Molecular Weight (g/mol) 224.27
Formula Weight 224.124 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.