missing translation for 'onlineSavingsMsg'
Learn More

Carisoprodol (1.0mg/ml in Methanol), TRC

Product Code. 18050940
Change view
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18050940 5 x 1 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18050940 Supplier Toronto Research Chemicals Supplier No. TRCKIT20205X1ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 78-44-4
Molecular Formula C12 H24 N2 O4
Quantity 5 x 1 mL
Synonym Carisoprodol, (2RS)-2-[(Carbamoyloxy)methyl]-2-methylpentyl (1-methylethyl)carbamate, Carbamic acid, N-(1-methylethyl)-, 2-[[(aminocarbonyl)oxy]methyl]-2-methylpentyl ester, Carbamic acid, (1-methylethyl)-, 2-[[(aminocarbonyl)oxy]methyl]-2-methylpentyl ester (9CI), Carbamic acid, isopropyl-, 2-(hydroxymethyl)-2-methylpentyl ester carbamate (ester) (8CI), Carbamic acid, isopropyl-, 2-(hydroxymethyl)-2-methylpentyl ester, carbamate (6CI), 2-Methyl-2-propyl-1,3-propanediol carbamate isopropylcarbamate, Apesan, Arusal, Atonalyt, Calenfa, Caprodat, Carisol, Carisoma, Carisoprodate, Carisoprodatum, Domarax, Flexal, Flexartal, Isobamate, Isomeprobamate, Isopropyl meprobamate, Isoprotan, Isoprotane, Isoprothane, Izoprotan, Miolisodal, Mioril, N-Isopropyl-2-methyl-2-propyl-1,3-propanediol dicarbamate, NIH 10966, NSC 172124, Rela, Relasom, Sanoma, Skutamil, Soma, Somadril, Somalgit, Stialgin
InChI Formula InChI=1S/C12H24N2O4/c1-5-6-12(4,7-17-10(13)15)8-18-11(16)14-9(2)3/h9H,5-8H2,1-4H3,(H2,13,15)(H,14,16)
SMILES CCCC(C)(COC(=O)N)COC(=O)NC(C)C
IUPAC Name [2-(carbamoyloxymethyl)-2-methylpentyl] N-propan-2-ylcarbamate
Molecular Weight (g/mol) 260.33
Formula Weight 260.1736
Concentration or Composition (by Analyte or Components) Carisoprodol 1 mg/mL
Chemical Name or Material Carisoprodol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.