missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Catalase Mouse anti-H. Pylori, Clone: HPYL/7170, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-13821-100UG
This item is not returnable.
View return policy
Description
Catalase Monoclonal antibody specifically detects Catalase in H. Pylori samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Catalase | |
| Monoclonal | |
| 0.2 mg/ml | |
| Immunohistochemistry-Paraffin 1-2 ug/ml | |
| catalase, EC 1.11.1.6, MGC138422, MGC138424 | |
| Recombinant Helicobacter pylori Catalase protein fragment (around aa323-445) (Exact sequence is proprietary) | |
| 100 μg | |
| Cancer, Cellular Markers, Hypoxia, Lipid and Metabolism, Neuroscience, Peroxisome Markers, Signal Transduction | |
| 847 | |
| Store at 4°C. | |
| IgG2b κ |
| Immunohistochemistry (Paraffin) | |
| HPYL/7170 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| H. pylori | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction