missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Catalase Mouse anti-H. Pylori, Clone: HPYL/7170, Novus Biologicals™
Mouse Monoclonal Antibody
250.00€ - 510.00€
Specifications
| Antigen | Catalase |
|---|---|
| Clone | HPYL/7170 |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
| Applications | Immunohistochemistry (Paraffin) |
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|
18366403
|
Novus Biologicals
NBP3-13821-20UG |
20 μg |
250.00€
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18311175
|
Novus Biologicals
NBP3-13821-100UG |
100 μg |
510.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
Catalase Monoclonal antibody specifically detects Catalase in H. Pylori samples. It is validated for Immunohistochemistry (Paraffin)Spezifikation
| Catalase | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Cellular Markers, Hypoxia, Lipid and Metabolism, Neuroscience, Peroxisome Markers, Signal Transduction | |
| 10 mM PBS with 0.05% BSA | |
| 847 | |
| IgG2b κ | |
| Protein A or G purified |
| HPYL/7170 | |
| Immunohistochemistry-Paraffin 1-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| H. pylori | |
| catalase, EC 1.11.1.6, MGC138422, MGC138424 | |
| Recombinant Helicobacter pylori Catalase protein fragment (around aa323-445) (Exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts